3-Bromo-4-chloro-2-hydroxybenzaldehyde 95%
Dodavatel: Apollo Scientific
OR315877-1G
OR315877-500MG
APOSOR315877-1GEA
29960
CZK
APOSOR315877-1G
APOSOR315877-500MG
3-Bromo-4-chloro-2-hydroxybenzaldehyde 95%
3-Bromo-4-chloro-2-hydroxybenzaldehyde
Vzorec:
C₇H₄BrClO₂ MW: 235,462 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23705580 CAS číslo: 872274-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...