Tisk…

2-Cyclopropylpyridine-3-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR315872-1G OR315872-250MG
APOSOR315872-1GEA 15510 CZK
APOSOR315872-1G APOSOR315872-250MG
2-Cyclopropylpyridine-3-carbaldehyde 95%
2-Cyclopropylpyridine-3-carbaldehyde
Vzorec: C₉H₉NO
Storage Temperature: Ambient
MDL Number: MFCD18255582
CAS číslo: 921760-70-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR