Tisk…

1-(5-Cyclopropylpyridin-2-yl)ethanone

Dodavatel: Apollo Scientific

OR315870-1G OR315870-250MG OR315870-5G
APOSOR315870-1GEA 13230 CZK
APOSOR315870-1G APOSOR315870-250MG APOSOR315870-5G
1-(5-Cyclopropylpyridin-2-yl)ethanone
1-(5-Cyclopropylpyridin-2-yl)ethanone
Vzorec: C₁₀H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD18254232
CAS číslo: 188918-75-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR