3-Cyclopropyl-4-(hydroxymethyl)thiophene
Dodavatel: Apollo Scientific
OR315858-1G
OR315858-250MG
APOSOR315858-1GEA
18620
CZK
APOSOR315858-1G
APOSOR315858-250MG
3-Cyclopropyl-4-(hydroxymethyl)thiophene
3-Cyclopropyl-4-(hydroxymethyl)thiophene
Vzorec:
C₈H₁₀OS MW: 154,229 g/mol Storage Temperature: Ambient |
CAS číslo:
1602792-09-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...