Tisk…

1-(3-Bromophenyl)-2-methylpropan-1-one 98%

Dodavatel: Apollo Scientific

OR315509-1G OR315509-5G
APOSOR315509-1GEA 5200 CZK
APOSOR315509-1G APOSOR315509-5G
1-(3-Bromophenyl)-2-methylpropan-1-one 98%
1-(3-Bromophenyl)-2-methylpropan-1-one
Vzorec: C₁₀H₁₁BrO
MW: 227,098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08437145
CAS číslo: 2415-93-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR