Tisk…

2,8-Diaza-spiro[4.5]decane-2,3,8-tricarboxylic acid 8-tert-butyl ester 2-(9H-fluoren-9-ylmethyl) ester 95+%

Dodavatel: Apollo Scientific

Synonyms: 2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-8-(tert-butoxycarbonyl)-2,8-diazaspiro[4.5]decane-3-carboxylic acid

OR315422-100MG OR315422-1G OR315422-250MG
APOSOR315422-100MGEA 14290 CZK
APOSOR315422-100MG APOSOR315422-1G APOSOR315422-250MG
2,8-Diaza-spiro[4.5]decane-2,3,8-tricarboxylic acid 8-tert-butyl ester 2-(9H-fluoren-9-ylmethyl) ester 95+%
2,8-Diaza-spiro[4.5]decane-2,3,8-tricarboxylic acid 8-tert-butyl ester 2-(9H-fluoren-9-ylmethyl) ester
Vzorec: C₂₉H₃₄N₂O₆
MW: 506.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779072
CAS číslo: 849928-23-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR