Tisk…

Ethyl 5-bromo-6-hydroxypicolinate 98%

Dodavatel: Apollo Scientific

OR315313-1G OR315313-100MG OR315313-250MG
APOSOR315313-1GEA 8600 CZK
APOSOR315313-1G APOSOR315313-100MG APOSOR315313-250MG
Ethyl 5-bromo-6-hydroxypicolinate 98%
Ethyl 5-bromo-6-hydroxypicolinate
Vzorec: C₈H₈BrNO₃
MW: 246,058 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698334
CAS číslo: 1214346-74-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR