Tisk…

N-(5-Bromothiazol-2-yl)acetamide hydrobromide 95+%

Dodavatel: Apollo Scientific

OR315192-1G OR315192-25G OR315192-5G
APOSOR315192-1GEA 5430 CZK
APOSOR315192-1G APOSOR315192-25G APOSOR315192-5G
N-(5-Bromothiazol-2-yl)acetamide hydrobromide 95+%
N-(5-Bromothiazol-2-yl)acetamide hydrobromide
Vzorec: C₅H₅BrN₂OS·BrH
Storage Temperature: Ambient
MDL Number: MFCD20921811
CAS číslo: 1354359-55-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR