Tisk…

2-Amino-2-methylpropanenitrile hydrochloride 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Amino-2-methylpropanenitrile HCl, 2-Amino-2-methylpropionitrile hydrochloride, alpha-Aminoisobutyronitrile hydrochloride

OR315172-25G OR315172-5G
APOSOR315172-25GEA 705 CZK
APOSOR315172-25G APOSOR315172-5G
2-Amino-2-methylpropanenitrile hydrochloride 95%
2-Amino-2-methylpropanenitrile hydrochloride
Vzorec: C₄H₈N₂·ClH
MW: 120,581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08456816
CAS číslo: 50846-36-1
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR