3-Bromo-6-iodo-2-methylpyridine
Dodavatel: Apollo Scientific
OR315109-1G
OR315109-250MG
OR315109-5G
APOSOR315109-1GEA
3870
CZK
APOSOR315109-1G
APOSOR315109-250MG
APOSOR315109-5G
3-Bromo-6-iodo-2-methylpyridine
3-Bromo-6-iodo-2-methylpyridine
Vzorec:
C₆H₅BrIN MW: 297,919 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11227150 CAS číslo: 1008361-77-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...