5-Bromo-2-chloro-3-methoxypyrazine
Dodavatel: Apollo Scientific
OR315095-1G
OR315095-250MG
OR315095-5G
APOSOR315095-1GEA
1740
CZK
APOSOR315095-1G
APOSOR315095-250MG
APOSOR315095-5G
5-Bromo-2-chloro-3-methoxypyrazine
5-Bromo-2-chloro-3-methoxypyrazine
Vzorec:
C₅H₄BrClN₂O MW: 223,455 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18250187 CAS číslo: 89283-94-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...