3-Bromo-2-methoxy-4-picoline
Dodavatel: Apollo Scientific
|
Varování
|
OR315079-1G
OR315079-5G
APOSOR315079-1GEA
1080
CZK
APOSOR315079-1G
APOSOR315079-5G
3-Bromo-2-methoxy-4-picoline
3-Bromo-2-methoxy-4-picoline
Vzorec:
C₇H₈BrNO Storage Temperature: Ambient |
MDL Number:
MFCD08061305 CAS číslo: 717843-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...