Tisk…

6-Chloro-4-iodo-3-hydroxypyridine

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Chloro-4-iodo-5-hydroxypyridine, 6-Chloro-4-iodopyridin-3-ol

OR314078-1G
APOSOR314078-1GEA 2200 CZK
APOSOR314078-1G
6-Chloro-4-iodo-3-hydroxypyridine
6-Chloro-4-iodo-3-hydroxypyridine
Vzorec: C₅H₃ClINO
MW: 255,441 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977445
CAS číslo: 877133-58-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR