3-Bromo-2-chloroaniline 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR313100-25G
APOSOR313100-25GEA
5260
CZK
APOSOR313100-25G
3-Bromo-2-chloroaniline 95%
3-Bromo-2-chloroaniline
Vzorec:
C₆H₅BrClN MW: 206,468 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11848536 CAS číslo: 56131-46-5 UN: 2019 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...