Tisk…

3-Amino-1-n-boc-azepane

Dodavatel: Apollo Scientific

OR313087-1G OR313087-5G
APOSOR313087-1GEA 4860 CZK
APOSOR313087-1G APOSOR313087-5G
3-Amino-1-n-boc-azepane
3-Amino-1-n-boc-azepane
Vzorec: C₁₁H₂₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06738754
CAS číslo: 609789-17-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR