Ethyl 3-oxotetrahydro-2H-pyran-4-carboxylate 95%
Dodavatel: Apollo Scientific
OR313076-1G
OR313076-5G
APOSOR313076-1GEA
9890
CZK
APOSOR313076-1G
APOSOR313076-5G
Ethyl 3-oxotetrahydro-2H-pyran-4-carboxylate 95%
Ethyl 3-oxotetrahydro-2H-pyran-4-carboxylate
Vzorec:
C₈H₁₂O₄ MW: 172,178 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19440853 CAS číslo: 388109-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...