Tisk…

5-Nitro-2,4,6-trichloropyrimidine

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Nitro-2,4,6-trichloro-1,3-diazine

OR313061-5G
APOSOR313061-5GEA 5960 CZK
APOSOR313061-5G
5-Nitro-2,4,6-trichloropyrimidine
5-Nitro-2,4,6-trichloropyrimidine
Vzorec: C₄Cl₃N₃O₂
MW: 228,421 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00091516
CAS číslo: 4359-87-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR