Tisk…

3-Methyl-1H-pyrrole-2-carboxylic acid

Dodavatel: Apollo Scientific

OR313040-1G OR313040-5G
APOSOR313040-1GEA 4200 CZK
APOSOR313040-1G APOSOR313040-5G
3-Methyl-1H-pyrrole-2-carboxylic acid
3-Methyl-1H-pyrrole-2-carboxylic acid
Vzorec: C₆H₇NO₂
MW: 125,125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07440095
CAS číslo: 90724-57-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR