Tisk…

N-Boc-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid

Dodavatel: Apollo Scientific

Synonyms: 2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid

OR313034-1G OR313034-250MG
APOSOR313034-1GEA 8240 CZK
APOSOR313034-1G APOSOR313034-250MG
N-Boc-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid
N-Boc-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid
Vzorec: C₁₅H₁₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD05861548
CAS číslo: 170097-67-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR