Tisk…

3-Amino-1-N-methyl-azetidine dihydrochloride 97%

Dodavatel: Apollo Scientific
Varování

OR313027-1G OR313027-5G
APOSOR313027-1GEA 9920 CZK
APOSOR313027-1G APOSOR313027-5G
3-Amino-1-N-methyl-azetidine dihydrochloride 97%
3-Amino-1-N-methyl-azetidine dihydrochloride
Vzorec: C₄H₁₀N₂·2ClH
MW: 159,057 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07366633
CAS číslo: 1139634-75-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR