Tisk…

Methyl (2R,3R,4S,5S)-4-(hydroxymethyl)cubane-1-carboxylate 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 4-(hydroxymethyl)cubane-1-carboxylate

OR312541-250MG OR312541-100MG
APOSOR312541-250MGEA 11740 CZK
APOSOR312541-250MG APOSOR312541-100MG
Methyl (2R,3R,4S,5S)-4-(hydroxymethyl)cubane-1-carboxylate 95%
Methyl (2R,3R,4S,5S)-4-(hydroxymethyl)cubane-1-carboxylate
Vzorec: C₁₁H₁₂O₃
MW: 192,211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458190
CAS číslo: 60462-19-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR