Tisk…

4-Cyanobicyclo[2.2.2]octane-1-carboxylic acid

Dodavatel: Apollo Scientific

OR312495-1G OR312495-250MG
APOSOR312495-1GEA 39680 CZK
APOSOR312495-1G APOSOR312495-250MG
4-Cyanobicyclo[2.2.2]octane-1-carboxylic acid
4-Cyanobicyclo[2.2.2]octane-1-carboxylic acid
Vzorec: C₁₀H₁₃NO₂
MW: 179,216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00967723
CAS číslo: 15941-09-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR