Tisk…

Bicyclo[2.2.2]octane-1,4-diyldimethanol

Dodavatel: Apollo Scientific

OR312493-1G OR312493-5G
APOSOR312493-1GEA 1410 CZK
APOSOR312493-1G APOSOR312493-5G
Bicyclo[2.2.2]octane-1,4-diyldimethanol
Bicyclo[2.2.2]octane-1,4-diyldimethanol
Vzorec: C₁₀H₁₈O₂
MW: 170,249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23135544
CAS číslo: 826-45-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR