Tisk…

Ethyl 3-iodobicyclo[1.1.1]pentane-1-carboxylate

Dodavatel: Apollo Scientific
Varování

OR312479-1G OR312479-250MG
APOSOR312479-1GEA 68440 CZK
APOSOR312479-1G APOSOR312479-250MG
Ethyl 3-iodobicyclo[1.1.1]pentane-1-carboxylate
Ethyl 3-iodobicyclo[1.1.1]pentane-1-carboxylate
Vzorec: C₈H₁₁IO₂
MW: 266.076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29037335
CAS číslo: 1936371-35-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR