Methyl 3-((tert-butoxycarbonyl)amino)bicyclo[1.1.1]pentane-1-carboxylate 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR312473-1G
OR312473-5G
APOSOR312473-1GEA
4370
CZK
APOSOR312473-1G
APOSOR312473-5G
Methyl 3-((tert-butoxycarbonyl)amino)bicyclo[1.1.1]pentane-1-carboxylate 97%
Methyl 3-((tert-butoxycarbonyl)amino)bicyclo[1.1.1]pentane-1-carboxylate
Vzorec:
C₁₂H₁₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD24476148 CAS číslo: 676371-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...