Tisk…

2-(Bicyclo[1.1.1]pentan-1-yl)acetic acid

Dodavatel: Apollo Scientific
Varování

OR312470-25MG OR312470-100MG
APOSOR312470-25MGEA 1160 CZK
APOSOR312470-25MG APOSOR312470-100MG
2-(Bicyclo[1.1.1]pentan-1-yl)acetic acid
2-(Bicyclo[1.1.1]pentan-1-yl)acetic acid
Vzorec: C₇H₁₀O₂
Storage Temperature: Refrigerator
MDL Number: MFCD19229645
CAS číslo: 131515-31-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR