3-Aminobicyclo[1.1.1]pentane-1-carboxylic acid
Dodavatel: Apollo Scientific
OR312458-1G
OR312458-250MG
APOSOR312458-1GEA
38690
CZK
APOSOR312458-1G
APOSOR312458-250MG
3-Aminobicyclo[1.1.1]pentane-1-carboxylic acid
3-Aminobicyclo[1.1.1]pentane-1-carboxylic acid
Vzorec:
C₆H₉NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD18826734 CAS číslo: 406947-32-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...