Tisk…

1-(Bromomethyl)bicyclo[1.1.1]pentane

Dodavatel: Apollo Scientific
Varování

OR312450-250MG OR312450-100MG
APOSOR312450-250MGEA 31080 CZK
APOSOR312450-250MG APOSOR312450-100MG
1-(Bromomethyl)bicyclo[1.1.1]pentane
1-(Bromomethyl)bicyclo[1.1.1]pentane
Vzorec: C₆H₉Br
Storage Temperature: Refrigerator
MDL Number: MFCD20633000
CAS číslo: 161043-38-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR