tert-Butyl (3-ethynylbicyclo[1.1.1]pentan-1-yl)carbamate 95%
Dodavatel: Apollo Scientific
OR312442-250MG
OR312442-100MG
APOSOR312442-250MGEA
45630
CZK
APOSOR312442-250MG
APOSOR312442-100MG
tert-Butyl (3-ethynylbicyclo[1.1.1]pentan-1-yl)carbamate 95%
tert-Butyl (3-ethynylbicyclo[1.1.1]pentan-1-yl)carbamate
Vzorec:
C₁₂H₁₇NO₂ MW: 207,269 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27987299 CAS číslo: 1638761-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...