Bicyclo[1.1.1]pentan-1-ylhydrazine dihydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
OR312411-1G
OR312411-250MG
OR312411-100MG
APOSOR312411-1GEA
66120
CZK
APOSOR312411-1G
APOSOR312411-250MG
APOSOR312411-100MG
Bicyclo[1.1.1]pentan-1-ylhydrazine dihydrochloride
Bicyclo[1.1.1]pentan-1-ylhydrazine dihydrochloride
Vzorec:
C₅H₁₀N₂·2ClH Storage Temperature: Refrigerator |
MDL Number:
MFCD27996126 CAS číslo: 1403746-38-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...