Tisk…

4-(Bicyclo[1.1.1]pentan-1-yl)aniline 95%

Dodavatel: Apollo Scientific
Varování

OR312394-1G OR312394-250MG
APOSOR312394-1GEA 68440 CZK
APOSOR312394-1G APOSOR312394-250MG
4-(Bicyclo[1.1.1]pentan-1-yl)aniline 95%
4-(Bicyclo[1.1.1]pentan-1-yl)aniline
Vzorec: C₁₁H₁₃N
Storage Temperature: Ambient
MDL Number: MFCD27996108
CAS číslo: 1823343-61-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR