Methyl 3-acetylbicyclo[1.1.1]pentane-1-carboxylate
Dodavatel: Apollo Scientific
OR312386-1G
OR312386-250MG
APOSOR312386-1GEA
31280
CZK
APOSOR312386-1G
APOSOR312386-250MG
Methyl 3-acetylbicyclo[1.1.1]pentane-1-carboxylate
Methyl 3-acetylbicyclo[1.1.1]pentane-1-carboxylate
Vzorec:
C₉H₁₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD24696091 CAS číslo: 131515-42-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...