Tisk…

3-Aminobicyclo[1.1.1]pentane-1-carboxylic acid, N-BOC protected 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl (3-carboxybicyclo[1.1.1]pent-1-yl)carbamate, 3-[(tert-Butoxycarbonyl)amino]bicyclo[1.1.1]pentane-1-carboxylic acid, 3-(Boc-amino)bicyclo[1.1.1]pentane-1-carboxylic Acid

OR312125-1G OR312125-5G
APOSOR312125-1GEA 4930 CZK
APOSOR312125-1G APOSOR312125-5G
3-Aminobicyclo[1.1.1]pentane-1-carboxylic acid, N-BOC protected 97%
3-Aminobicyclo[1.1.1]pentane-1-carboxylic acid, N-BOC protected
Vzorec: C₁₁H₁₇NO₄
MW: 227,257 g/mol
Bod tání: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD09971715
CAS číslo: 303752-38-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR