Tisk…

Bicyclo[1.1.1]pentane-1,3-dicarboxylic acid 98%

Dodavatel: Apollo Scientific
Varování

OR312111-1G OR312111-5G
APOSOR312111-1GEA 2030 CZK
APOSOR312111-1G APOSOR312111-5G
Bicyclo[1.1.1]pentane-1,3-dicarboxylic acid 98%
Bicyclo[1.1.1]pentane-1,3-dicarboxylic acid
Vzorec: C₇H₈O₄
Storage Temperature: Ambient
MDL Number: MFCD20621127
CAS číslo: 56842-95-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR