Tisk…

4-Bromo-1-(4-chlorophenyl)butan-1-one

Dodavatel: Apollo Scientific

OR311105-1G OR311105-500MG
APOSOR311105-1GEA 19110 CZK
APOSOR311105-1G APOSOR311105-500MG
4-Bromo-1-(4-chlorophenyl)butan-1-one
4-Bromo-1-(4-chlorophenyl)butan-1-one
Vzorec: C₁₀H₁₀BrClO
MW: 261,543 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16333877
CAS číslo: 156092-12-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR