Tisk…

Ethyl 4-(carbamoylamino)benzoate

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 4-ureidobenzoate

OR311007-1G OR311007-500MG OR311007-5G
APOSOR311007-1GEA 4300 CZK
APOSOR311007-1G APOSOR311007-500MG APOSOR311007-5G
Ethyl 4-(carbamoylamino)benzoate
Ethyl 4-(carbamoylamino)benzoate
Vzorec: C₁₀H₁₂N₂O₃
MW: 208.214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02379956
CAS číslo: 13289-38-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR