Tisk…

Ethyl 6-(chloromethyl)-4-(4-methoxyphenyl)-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate

Dodavatel: Apollo Scientific

OR310997-1G
APOSOR310997-1GEA 12670 CZK
APOSOR310997-1G
Ethyl 6-(chloromethyl)-4-(4-methoxyphenyl)-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Ethyl 6-(chloromethyl)-4-(4-methoxyphenyl)-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Vzorec: C₁₅H₁₇ClN₂O₄
MW: 324,759 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00749776
CAS číslo: 475042-38-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR