4-{[3-(4-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]methyl}piperidine hydrochloride
Dodavatel: Apollo Scientific
OR310994-1G
APOSOR310994-1GEA
22220
CZK
APOSOR310994-1G
4-{[3-(4-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]methyl}piperidine hydrochloride
4-{[3-(4-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]methyl}piperidine hydrochloride
Vzorec:
C₁₅H₁₉N₃O₂·ClH MW: 309,791 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28041319 CAS číslo: 1622069-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...