Tisk…

2-(4-Methoxynaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Dodavatel: Apollo Scientific

Synonyms: 4-Methoxynaphthalene-1-boronic acid pinacol ester

OR310979-1G OR310979-5G
APOSOR310979-1GEA 4300 CZK
APOSOR310979-1G APOSOR310979-5G
2-(4-Methoxynaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-(4-Methoxynaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Vzorec: C₁₇H₂₁BO₃
MW: 284,158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414634
CAS číslo: 269410-16-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR