(4-Vinylphenyl)boronic acid pinacol ester
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4,4,5,5-Tetramethyl-2-(4-vinylphenyl)-1,3,2-dioxaborolane
OR310973-1G
OR310973-5G
APOSOR310973-1GEA
1780
CZK
APOSOR310973-1G
APOSOR310973-5G
(4-Vinylphenyl)boronic acid pinacol ester
(4-Vinylphenyl)boronic acid pinacol ester
Vzorec:
C₁₄H₁₉BO₂ MW: 230,11 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18733870 CAS číslo: 870004-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...