Tisk…
(2S)-5-Methoxy-1,2,3,4-tetrahydronaphthalen-2-amine hydrochloride
Dodavatel: Apollo Scientific
OR310968-1G OR310968-500MG
APOSOR310968-1GEA 9560 CZK
APOSOR310968-1G APOSOR310968-500MG
(2S)-5-Methoxy-1,2,3,4-tetrahydronaphthalen-2-amine hydrochloride
(2S)-5-Methoxy-1,2,3,4-tetrahydronaphthalen-2-amine hydrochloride
Vzorec: C₁₁H₁₅NO·ClH
MW: 213,704 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08443937
CAS číslo: 58349-17-0