Tisk…

8-Benzyl-8-azabicyclo[3.2.1]octan-3-one hydrochloride

Dodavatel: Apollo Scientific
Varování

OR310941-1G OR310941-5G
APOSOR310941-1GEA 622 CZK
APOSOR310941-1G APOSOR310941-5G
8-Benzyl-8-azabicyclo[3.2.1]octan-3-one hydrochloride
8-Benzyl-8-azabicyclo[3.2.1]octan-3-one hydrochloride
Vzorec: C₁₄H₁₇NO·ClH
MW: 251,752 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659471
CAS číslo: 83393-23-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR