Tisk…

5-(Benzyloxy)-4-oxo-4H-pyran-2-carboxylic acid

Dodavatel: Apollo Scientific

OR310900-1G OR310900-500MG
APOSOR310900-1GEA 9560 CZK
APOSOR310900-1G APOSOR310900-500MG
5-(Benzyloxy)-4-oxo-4H-pyran-2-carboxylic acid
5-(Benzyloxy)-4-oxo-4H-pyran-2-carboxylic acid
Vzorec: C₁₃H₁₀O₅
MW: 246,216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08531038
CAS číslo: 1219-33-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR