5-Bromopyrimidine-2,4-diamine
Dodavatel: Apollo Scientific
OR310886-1G
OR310886-500MG
APOSOR310886-1GEA
25460
CZK
APOSOR310886-1G
APOSOR310886-500MG
5-Bromopyrimidine-2,4-diamine
5-Bromopyrimidine-2,4-diamine
Vzorec:
C₄H₅BrN₄ MW: 189,013 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11848482 CAS číslo: 1004-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...