Tisk…

2-Bromo-5-methoxy-3-nitropyridine

Dodavatel: Apollo Scientific
Varování

OR31051-1G OR31051-250MG
APOSOR31051-1GEA 12140 CZK
APOSOR31051-1G APOSOR31051-250MG
2-Bromo-5-methoxy-3-nitropyridine
2-Bromo-5-methoxy-3-nitropyridine
Vzorec: C₆H₅BrN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD21605255
CAS číslo: 1805025-96-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR