Tisk…

N-Cyclopropyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

Dodavatel: Apollo Scientific

OR310459-1G OR310459-500MG
APOSOR310459-1GEA 6780 CZK
APOSOR310459-1G APOSOR310459-500MG
N-Cyclopropyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
N-Cyclopropyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Vzorec: C₁₄H₂₁BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09027072
CAS číslo: 2096334-77-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR