2-[4-(3-Amino-4-nitrophenyl)piperazin-1-yl]ethan-1-ol
Dodavatel: Apollo Scientific
OR310457-5G
OR310457-1G
APOSOR310457-5GEA
13330
CZK
APOSOR310457-5G
APOSOR310457-1G
2-[4-(3-Amino-4-nitrophenyl)piperazin-1-yl]ethan-1-ol
2-[4-(3-Amino-4-nitrophenyl)piperazin-1-yl]ethan-1-ol
Vzorec:
C₁₂H₁₈N₄O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00545981 CAS číslo: 23470-44-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...