Tisk…

Phenyl N-hydroxycarbamate

Dodavatel: Apollo Scientific
Varování

OR310437-1G OR310437-5G OR310437-10G
APOSOR310437-1GEA 5130 CZK
APOSOR310437-1G APOSOR310437-5G APOSOR310437-10G
Phenyl N-hydroxycarbamate
Phenyl N-hydroxycarbamate
Vzorec: C₇H₇NO₃
MW: 153,135 g/mol
Bod tání: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD18968936
CAS číslo: 38064-07-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR