Tisk…

Benzyl N-[(1R)-1-(hydrazinecarbonyl)ethyl]carbamate

Dodavatel: Apollo Scientific

OR310382-1G OR310382-500MG
APOSOR310382-1GEA 19110 CZK
APOSOR310382-1G APOSOR310382-500MG
Benzyl N-[(1R)-1-(hydrazinecarbonyl)ethyl]carbamate
Benzyl N-[(1R)-1-(hydrazinecarbonyl)ethyl]carbamate
Vzorec: C₁₁H₁₅N₃O₃
MW: 237,255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26936353
CAS číslo: 57355-13-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR