3-Bromo-2-(2-chlorophenoxy)pyridine
Dodavatel: Apollo Scientific
OR310376-1G
OR310376-500MG
APOSOR310376-1GEA
19110
CZK
APOSOR310376-1G
APOSOR310376-500MG
3-Bromo-2-(2-chlorophenoxy)pyridine
3-Bromo-2-(2-chlorophenoxy)pyridine
Vzorec:
C₁₁H₇BrClNO MW: 284,536 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14651748 CAS číslo: 1215809-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...